Product Details

SNP ID
rs140969766
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55823115 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTACACATCATAGTATTCATTGCC[G/T]CCTTGAGGTCATCTTGCAGCTTGGA
Phenotype
MIM: 606092 MIM: 610074 MIM: 610049
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DNAJC14 PubMed Links

Gene Details

Gene
DNAJC14
Gene Name
DnaJ heat shock protein family (Hsp40) member C14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032364.5 1793 Missense Mutation GAG,GCG E530A NP_115740.5
Gene
ORMDL2
Gene Name
ORMDL sphingolipid biosynthesis regulator 2
There are no transcripts associated with this gene.

Gene
SARNP
Gene Name
SAP domain containing ribonucleoprotein
There are no transcripts associated with this gene.

Gene
TMEM198B
Gene Name
transmembrane protein 198B (pseudogene)
There are no transcripts associated with this gene.

View Full Product Details