Product Details

SNP ID
rs141145771
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:48330017 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGAAGCTCCAAGCCCAGGGAAGA[C/G]AAGCAGGAGCCCAAGAAGCCCGCAC
Phenotype
MIM: 613906
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
H1FNT PubMed Links

Gene Details

Gene
H1FNT
Gene Name
H1 histone family member N, testis specific
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181788.1 1038 Missense Mutation GAC,GAG D242E NP_861453.1
Gene
ZNF641
Gene Name
zinc finger protein 641
There are no transcripts associated with this gene.

View Full Product Details