Product Details

SNP ID
rs141179863
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:48691022 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCTACCAATTATCTAACACAACA[C/T]AGAAAATATCAAGGGGAAAGAGGAA
Phenotype
MIM: 143055 MIM: 615488
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCNT1 PubMed Links

Gene Details

Gene
CCNT1
Gene Name
cyclin T1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001240.3 4351 UTR 3 NP_001231.2
NM_001277842.1 4351 UTR 3 NP_001264771.1
XM_017020197.1 4351 UTR 3 XP_016875686.1
Gene
KANSL2
Gene Name
KAT8 regulatory NSL complex subunit 2
There are no transcripts associated with this gene.

View Full Product Details