Product Details

SNP ID
rs141404931
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:122867191 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGAGGGAGAGGTGGCGGATATGG[A/G]GACACGAACTGCGAAGAGAATCCTT
Phenotype
MIM: 605613 MIM: 610037
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HIP1R PubMed Links

Gene Details

Gene
HIP1R
Gene Name
huntingtin interacting protein 1 related
There are no transcripts associated with this gene.

Gene
VPS37B
Gene Name
VPS37B, ESCRT-I subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024667.2 886 Silent Mutation TCC,TCT S261S NP_078943.1
XM_005253622.3 886 Silent Mutation TCC,TCT S233S XP_005253679.1
XM_006719602.3 886 Silent Mutation TCC,TCT S233S XP_006719665.1
XM_011538743.2 886 Silent Mutation TCC,TCT S320S XP_011537045.1
XM_011538745.2 886 Silent Mutation TCC,TCT S243S XP_011537047.1
XM_017019970.1 886 Intron XP_016875459.1
XM_017019971.1 886 Intron XP_016875460.1
XM_017019972.1 886 Intron XP_016875461.1

View Full Product Details