Product Details

SNP ID
rs141647931
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:76346148 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTTTTTGGCATAATTGAGAAGAT[A/G]GTAATGTAACAAGATCTCAAAATTA
Phenotype
MIM: 610148 MIM: 606736
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BBS10 PubMed Links

Gene Details

Gene
BBS10
Gene Name
Bardet-Biedl syndrome 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024685.3 1921 Missense Mutation CAT,TAT H613Y NP_078961.3
Gene
OSBPL8
Gene Name
oxysterol binding protein like 8
There are no transcripts associated with this gene.

View Full Product Details