Product Details

SNP ID
rs141661593
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:108623475 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGGGTATGAACAGACCTCTTTTG[A/G]TAGTAGGTTCCATGGACAGGGCCTC
Phenotype
MIM: 600738
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SELPLG PubMed Links

Gene Details

Gene
SELPLG
Gene Name
selectin P ligand
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206609.1 1058 Missense Mutation ACC,ATC T294I NP_001193538.1
NM_003006.4 1058 Missense Mutation ACC,ATC T278I NP_002997.2

View Full Product Details