Product Details

SNP ID
rs141743480
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:77024180 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATGCTCTTGGGGGTCACTGGAAC[G/T]GGTGACTGAAAAAAGAAAAAAGAAA
Phenotype
MIM: 612046
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
E2F7 PubMed Links

Gene Details

Gene
E2F7
Gene Name
E2F transcription factor 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203394.2 2807 Silent Mutation CCA,CCC P857P NP_976328.2
XM_011537966.2 2807 Silent Mutation CCA,CCC P812P XP_011536268.1
XM_011537969.2 2807 Silent Mutation CCA,CCC P756P XP_011536271.1

View Full Product Details