Product Details

SNP ID
rs142004064
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:4543506 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAATCATTTTTTCTTTGACACTTA[C/T]ATGCTGTCTTAATTGCTTTTGCAGT
Phenotype
MIM: 616082 MIM: 603070
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C12orf4 PubMed Links

Gene Details

Gene
C12orf4
Gene Name
chromosome 12 open reading frame 4
There are no transcripts associated with this gene.

Gene
RAD51AP1
Gene Name
RAD51 associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130862.1 Intron NP_001124334.1
NM_006479.4 Intron NP_006470.1

View Full Product Details