Product Details

SNP ID
rs142230441
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:98732501 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTAAGAAAATACACGTGTCCCCT[C/G]ACTTCAAAACATATGTGACTGTGGA
Phenotype
MIM: 607815 MIM: 602233
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ANKS1B PubMed Links

Gene Details

Gene
ANKS1B
Gene Name
ankyrin repeat and sterile alpha motif domain containing 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204065.1 7368 Intron NP_001190994.1
NM_001204066.1 7368 Intron NP_001190995.1
NM_001204067.1 7368 Intron NP_001190996.1
NM_001204068.1 7368 Intron NP_001190997.1
NM_001204069.1 7368 Intron NP_001190998.1
NM_001204070.1 7368 Intron NP_001190999.1
NM_001204079.1 7368 Intron NP_001191008.1
NM_001204080.1 7368 Intron NP_001191009.1
NM_001204081.1 7368 Intron NP_001191010.1
NM_020140.3 7368 Intron NP_064525.1
NM_152788.4 7368 Intron NP_690001.3
NM_181670.3 7368 Intron NP_858056.2
XM_005269028.4 7368 Intron XP_005269085.1
XM_005269029.4 7368 Intron XP_005269086.1
XM_005269032.3 7368 Intron XP_005269089.1
XM_006719504.3 7368 Intron XP_006719567.1
XM_006719505.3 7368 Intron XP_006719568.1
XM_006719506.3 7368 Intron XP_006719569.1
XM_006719507.3 7368 Intron XP_006719570.1
XM_006719508.3 7368 Intron XP_006719571.1
XM_006719509.3 7368 Intron XP_006719572.1
XM_006719510.3 7368 Intron XP_006719573.1
XM_006719512.3 7368 Intron XP_006719575.1
XM_006719513.3 7368 Intron XP_006719576.1
XM_006719514.3 7368 Intron XP_006719577.1
XM_011538571.2 7368 Intron XP_011536873.1
XM_017019651.1 7368 Intron XP_016875140.1
XM_017019652.1 7368 Intron XP_016875141.1
XM_017019653.1 7368 UTR 3 XP_016875142.1
XM_017019654.1 7368 Intron XP_016875143.1
XM_017019655.1 7368 Intron XP_016875144.1
XM_017019656.1 7368 Intron XP_016875145.1
XM_017019657.1 7368 Intron XP_016875146.1
XM_017019658.1 7368 Intron XP_016875147.1
XM_017019659.1 7368 Intron XP_016875148.1
XM_017019660.1 7368 Intron XP_016875149.1
XM_017019661.1 7368 Intron XP_016875150.1
XM_017019662.1 7368 Intron XP_016875151.1
XM_017019663.1 7368 Intron XP_016875152.1
XM_017019664.1 7368 Intron XP_016875153.1
XM_017019665.1 7368 Intron XP_016875154.1
Gene
APAF1
Gene Name
apoptotic peptidase activating factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160.2 7368 Missense Mutation CAC,GAC H1174D NP_001151.1
NM_013229.2 7368 Missense Mutation CAC,GAC H1217D NP_037361.1
NM_181861.1 7368 Missense Mutation CAC,GAC H1228D NP_863651.1
NM_181868.1 7368 Missense Mutation CAC,GAC H1185D NP_863658.1
NM_181869.1 7368 UTR 3 NP_863659.1
XM_017019250.1 7368 Intron XP_016874739.1

View Full Product Details