Product Details

SNP ID
rs143001555
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:15918049 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCACACTAATACCACCAGTTCTG[A/G]TCCAACACCACAGGATTTATCCCAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DERA PubMed Links

Gene Details

Gene
DERA
Gene Name
deoxyribose-phosphate aldolase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300779.1 Intron NP_001287708.1
NM_015954.3 Intron NP_057038.2

View Full Product Details