Product Details

SNP ID
rs143166654
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56131090 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCACCCAGGTCAGAGAAAAGAGCA[G/T]ATCCTGCTGGACTTGAACATCAGGT
Phenotype
MIM: 616670
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ESYT1 PubMed Links

Gene Details

Gene
ESYT1
Gene Name
extended synaptotagmin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184796.1 736 Missense Mutation CAG,CAT Q206H NP_001171725.1
NM_015292.2 736 Missense Mutation CAG,CAT Q206H NP_056107.1
Gene
ZC3H10
Gene Name
zinc finger CCCH-type containing 10
There are no transcripts associated with this gene.

View Full Product Details