Product Details
- SNP ID
-
rs143478289
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:52394788 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGGTCTGCACACAGGTGAGTGGAA[G/T]GTGACTCATGAGGCAAAGGAGGGGT
- Phenotype
-
MIM: 601078
MIM: 602766
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
KRT82
PubMed Links
Gene Details
- Gene
- KRT82
- Gene Name
- keratin 82
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_033033.3 |
1844 |
UTR 3 |
|
|
NP_149022.3 |
- Gene
- KRT84
- Gene Name
- keratin 84
There are no transcripts associated with this gene.
View Full Product Details