Product Details

SNP ID
rs143604047
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:123585373 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCTACTCTTTACGCCTCCAGCTA[C/T]CTCGTTTAGTCTCTTCTGCCAAGGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101927415 PubMed Links

Gene Details

Gene
LOC101927415
Gene Name
uncharacterized LOC101927415
There are no transcripts associated with this gene.

Gene
TMED2
Gene Name
transmembrane p24 trafficking protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321445.1 Intron NP_001308374.1
NM_006815.3 Intron NP_006806.1

View Full Product Details