Product Details

SNP ID
rs143731393
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:119593431 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATTAAGTGGCTTTTAAAAATGAG[C/T]TGGTCATCTTCAGCCCACGGGCTGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105370027 PubMed Links

Gene Details

Gene
LOC105370027
Gene Name
uncharacterized LOC105370027
There are no transcripts associated with this gene.

Gene
TMEM233
Gene Name
transmembrane protein 233
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136534.1 1309 Intron NP_001130006.1
XM_005253880.3 1309 UTR 5 XP_005253937.1
XM_005253881.3 1309 UTR 5 XP_005253938.1
XM_011538328.2 1309 UTR 5 XP_011536630.1
XM_011538329.2 1309 UTR 5 XP_011536631.1
XM_011538330.2 1309 UTR 5 XP_011536632.1
XM_011538331.2 1309 UTR 5 XP_011536633.1

View Full Product Details