Product Details

SNP ID
rs144082835
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:48202978 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCACTCGGCCACACCAACACAGAA[C/G]ACAAAGTACATCTGGGTCATGCAAC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR10AD1 PubMed Links

Gene Details

Gene
OR10AD1
Gene Name
olfactory receptor family 10 subfamily AD member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004134.1 315 Silent Mutation GTC,GTG V105V NP_001004134.1

View Full Product Details