Product Details

SNP ID
rs144105987
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:122235893 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGGCCGGCCTTCTCCAGGTTGT[G/T]TAAGGTCAATATGTGCTCATAGCCG
Phenotype
MIM: 605219 MIM: 610034
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DIABLO PubMed Links

Gene Details

Gene
DIABLO
Gene Name
diablo IAP-binding mitochondrial protein
There are no transcripts associated with this gene.

Gene
LOC101593348
Gene Name
uncharacterized LOC101593348
There are no transcripts associated with this gene.

Gene
VPS33A
Gene Name
VPS33A, CORVET/HOPS core subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022916.4 1446 Missense Mutation AAC,CAC N445H NP_075067.2

View Full Product Details