Product Details

SNP ID
rs144260932
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56595824 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACAGAGGGTATGGAACAGGAGCC[C/T]CTGTTGTTCCCTTGCCTGTGGTCTC
Phenotype
MIM: 605682 MIM: 602387
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BAZ2A PubMed Links

Gene Details

Gene
BAZ2A
Gene Name
bromodomain adjacent to zinc finger domain 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300905.1 8114 UTR 3 NP_001287834.1
NM_013449.3 8114 UTR 3 NP_038477.2
XM_011537797.2 8114 Intron XP_011536099.1
XM_011537798.2 8114 Intron XP_011536100.1
XM_011537799.2 8114 Intron XP_011536101.1
XM_011537800.2 8114 Intron XP_011536102.1
XM_011537801.2 8114 Intron XP_011536103.1
XM_011537803.2 8114 Intron XP_011536105.1
XM_011537804.2 8114 Intron XP_011536106.1
XM_011537805.2 8114 Intron XP_011536107.1
XM_011537806.2 8114 Intron XP_011536108.1
XM_011537807.2 8114 Intron XP_011536109.1
XM_011537809.2 8114 Intron XP_011536111.1
XM_017018740.1 8114 Intron XP_016874229.1
XM_017018741.1 8114 Intron XP_016874230.1
XM_017018742.1 8114 Intron XP_016874231.1
Gene
RBMS2
Gene Name
RNA binding motif single stranded interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002898.3 8114 UTR 3 NP_002889.1
XM_005269059.4 8114 Intron XP_005269116.1
XM_005269060.4 8114 Intron XP_005269117.1
XM_005269061.3 8114 Intron XP_005269118.1
XM_005269066.4 8114 Intron XP_005269123.1
XM_006719541.3 8114 Intron XP_006719604.1
XM_006719542.3 8114 Intron XP_006719605.1
XM_006719543.3 8114 Intron XP_006719606.1
XM_006719544.3 8114 Intron XP_006719607.1
XM_011538637.2 8114 Intron XP_011536939.1
XM_011538638.2 8114 Intron XP_011536940.1
XM_011538639.2 8114 Intron XP_011536941.1
XM_011538640.2 8114 Intron XP_011536942.1
XM_011538642.2 8114 Intron XP_011536944.1
XM_017019775.1 8114 Intron XP_016875264.1
XM_017019776.1 8114 Intron XP_016875265.1
XM_017019777.1 8114 Intron XP_016875266.1
XM_017019778.1 8114 Intron XP_016875267.1

View Full Product Details