Product Details

SNP ID
rs144517803
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56596545 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAACCATGGAGATTTCACTCCCC[C/G]ATCCACCTCACAATTTCACTCCCCC
Phenotype
MIM: 605682 MIM: 602387
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BAZ2A PubMed Links

Gene Details

Gene
BAZ2A
Gene Name
bromodomain adjacent to zinc finger domain 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300905.1 7994 UTR 3 NP_001287834.1
NM_013449.3 7994 UTR 3 NP_038477.2
XM_011537797.2 7994 Intron XP_011536099.1
XM_011537798.2 7994 Intron XP_011536100.1
XM_011537799.2 7994 Intron XP_011536101.1
XM_011537800.2 7994 Intron XP_011536102.1
XM_011537801.2 7994 Intron XP_011536103.1
XM_011537803.2 7994 Intron XP_011536105.1
XM_011537804.2 7994 Intron XP_011536106.1
XM_011537805.2 7994 Intron XP_011536107.1
XM_011537806.2 7994 Intron XP_011536108.1
XM_011537807.2 7994 Intron XP_011536109.1
XM_011537809.2 7994 Intron XP_011536111.1
XM_017018740.1 7994 Intron XP_016874229.1
XM_017018741.1 7994 Intron XP_016874230.1
XM_017018742.1 7994 Intron XP_016874231.1
Gene
RBMS2
Gene Name
RNA binding motif single stranded interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002898.3 7994 Intron NP_002889.1
XM_005269059.4 7994 Intron XP_005269116.1
XM_005269060.4 7994 Intron XP_005269117.1
XM_005269061.3 7994 Intron XP_005269118.1
XM_005269066.4 7994 Intron XP_005269123.1
XM_006719541.3 7994 Intron XP_006719604.1
XM_006719542.3 7994 Intron XP_006719605.1
XM_006719543.3 7994 Intron XP_006719606.1
XM_006719544.3 7994 Intron XP_006719607.1
XM_011538637.2 7994 Intron XP_011536939.1
XM_011538638.2 7994 Intron XP_011536940.1
XM_011538639.2 7994 Intron XP_011536941.1
XM_011538640.2 7994 Intron XP_011536942.1
XM_011538642.2 7994 Intron XP_011536944.1
XM_017019775.1 7994 Intron XP_016875264.1
XM_017019776.1 7994 Intron XP_016875265.1
XM_017019777.1 7994 Intron XP_016875266.1
XM_017019778.1 7994 Intron XP_016875267.1

View Full Product Details