Product Details

SNP ID
rs144640797
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:10620250 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTACTTGTAACACAGCCTCGTCA[C/T]CTGCAGTTTTAATGGCAGCTTCTAG
Phenotype
MIM: 611433
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAGOHB PubMed Links

Gene Details

Gene
MAGOHB
Gene Name
mago homolog B, exon junction complex core component
There are no transcripts associated with this gene.

Gene
STYK1
Gene Name
serine/threonine/tyrosine kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018423.2 1684 Missense Mutation GAT,GGT D388G NP_060893.2
XM_005253417.2 1684 Missense Mutation GAT,GGT D388G XP_005253474.1
XM_011520736.1 1684 Missense Mutation GAT,GGT D388G XP_011519038.1
XM_011520737.2 1684 Missense Mutation GAT,GGT D388G XP_011519039.1
XM_011520738.2 1684 Missense Mutation GAT,GGT D388G XP_011519040.1
XM_017019579.1 1684 Missense Mutation GAT,GGT D256G XP_016875068.1

View Full Product Details