Product Details

SNP ID
rs144644143
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:132844056 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACTCGGAAGCAGGAATGTTCTGC[C/T]GATACTGATAGGTCAGCTCACGGAA
Phenotype
MIM: 605209
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHFR PubMed Links

Gene Details

Gene
CHFR
Gene Name
checkpoint with forkhead and ring finger domains, E3 ubiquitin protein ligase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161344.1 1811 Missense Mutation CAG,CGG Q617R NP_001154816.1
NM_001161345.1 1811 Missense Mutation CAG,CGG Q616R NP_001154817.1
NM_001161346.1 1811 Missense Mutation CAG,CGG Q605R NP_001154818.1
NM_001161347.1 1811 Missense Mutation CAG,CGG Q525R NP_001154819.1
NM_018223.2 1811 Missense Mutation CAG,CGG Q576R NP_060693.2

View Full Product Details