Product Details

SNP ID
rs144968885
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:75498701 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCCCCTAACTTTACAGTTAACCG[A/T]CAGCGAGACCAAGTCAAACGTACGT
Phenotype
MIM: 602692 MIM: 612817
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GLIPR1 PubMed Links

Gene Details

Gene
GLIPR1
Gene Name
GLI pathogenesis related 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006851.2 825 Silent Mutation CGA,CGT R209R NP_006842.2
XM_011537775.2 825 Missense Mutation GAC,GTC D233V XP_011536077.1
XM_017018721.1 825 Missense Mutation GAC,GTC D92V XP_016874210.1
Gene
KRR1
Gene Name
KRR1, small subunit processome component homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007043.6 825 UTR 3 NP_008974.5

View Full Product Details