Product Details

SNP ID
rs145131024
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:49094213 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTAGGGAGAGGCCCTCCTTACCTC[A/G]GTCATCAGGCGGTCGGCTCCACTGT
Phenotype
MIM: 605423 MIM: 610007
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DHH PubMed Links

Gene Details

Gene
DHH
Gene Name
desert hedgehog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021044.3 607 Silent Mutation NP_066382.1
XM_017019380.1 607 Intron XP_016874869.1
XM_017019381.1 607 Intron XP_016874870.1
Gene
LMBR1L
Gene Name
limb development membrane protein 1 like
There are no transcripts associated with this gene.

View Full Product Details