Product Details

SNP ID
rs145384583
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55492650 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCAACTGGGAACAAAATAATAAC[A/G]TATAATGCATGTGTCATTCAGCTAT
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
OR6C68 PubMed Links

Gene Details

Gene
OR6C68
Gene Name
olfactory receptor family 6 subfamily C member 68
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005519.2 273 Silent Mutation ACA,ACG T91T NP_001005519.2

View Full Product Details