Product Details

SNP ID
rs145514203
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:108623248 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTCGGTGGGGGAGTAATTACGCA[C/T]GGGGTACATGTGGCCCTTGCGGGAG
Phenotype
MIM: 600738
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SELPLG PubMed Links

Gene Details

Gene
SELPLG
Gene Name
selectin P ligand
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206609.1 1285 Missense Mutation NP_001193538.1
NM_003006.4 1285 Missense Mutation NP_002997.2

View Full Product Details