Product Details

SNP ID
rs145537321
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55637284 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATATGTTGCCATCTGCCAGCCCCT[A/G]CGCTATTCCACCCTCTTGAGCCCAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR10P1 PubMed Links

Gene Details

Gene
OR10P1
Gene Name
olfactory receptor family 10 subfamily P member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_206899.1 393 Silent Mutation CTA,CTG L131L NP_996782.1

View Full Product Details