Product Details

SNP ID
rs145706089
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:111363250 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGTGCAGCCCTCCAGGATGATGA[C/T]GCCCACGGGCTCACGGCTGGCAGCG
Phenotype
MIM: 614239
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM109A PubMed Links

Gene Details

Gene
FAM109A
Gene Name
family with sequence similarity 109 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001177996.1 569 Missense Mutation ATC,GTC I73V NP_001171467.1
NM_001177997.1 569 Missense Mutation ATC,GTC I60V NP_001171468.1
NM_144671.4 569 Missense Mutation ATC,GTC I60V NP_653272.2
XM_006719257.3 569 Missense Mutation ATC,GTC I60V XP_006719320.1
XM_011537976.2 569 Missense Mutation ATC,GTC I60V XP_011536278.1
XM_017018883.1 569 Missense Mutation ATC,GTC I60V XP_016874372.1

View Full Product Details