Product Details

SNP ID
rs145874174
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:112013474 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAGGAACCCGGCGATATGGCTGC[C/G]GCTGTGCCCCGCGCCGCATTTCTCT
Phenotype
MIM: 602287
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ERP29 PubMed Links

Gene Details

Gene
ERP29
Gene Name
endoplasmic reticulum protein 29
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001034025.1 127 Silent Mutation GCC,GCT A3A NP_001029197.1
NM_006817.3 127 Silent Mutation GCC,GCT A3A NP_006808.1
XM_017018720.1 127 Intron XP_016874209.1
Gene
TMEM116
Gene Name
transmembrane protein 116
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193453.1 127 Intron NP_001180382.1
NM_001193531.1 127 Intron NP_001180460.1
NM_001294314.1 127 Intron NP_001281243.1
NM_138341.2 127 Intron NP_612350.1
XM_006719678.3 127 Intron XP_006719741.1
XM_011538946.1 127 Intron XP_011537248.1
XM_011538947.1 127 Intron XP_011537249.1
XM_011538948.2 127 Intron XP_011537250.1
XM_011538949.2 127 Intron XP_011537251.1
XM_011538950.1 127 Intron XP_011537252.1
XM_011538951.2 127 Intron XP_011537253.1
XM_011538953.1 127 Intron XP_011537255.1
XM_011538955.1 127 Intron XP_011537257.1
XM_011538957.1 127 Intron XP_011537259.1
XM_011538958.1 127 Intron XP_011537260.1
XM_017020187.1 127 Intron XP_016875676.1
XM_017020188.1 127 Intron XP_016875677.1
XM_017020189.1 127 Intron XP_016875678.1
XM_017020190.1 127 Intron XP_016875679.1
XM_017020191.1 127 Intron XP_016875680.1
XM_017020192.1 127 Intron XP_016875681.1
XM_017020193.1 127 Intron XP_016875682.1
XM_017020194.1 127 Intron XP_016875683.1

View Full Product Details