Product Details

SNP ID
rs146204253
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:101880993 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTCTTTTAGGTTGTTTGCTCTCT[A/G]CTTACCTGAGCAGCCGGAGCACTGG
Phenotype
MIM: 610776
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DRAM1 PubMed Links

Gene Details

Gene
DRAM1
Gene Name
DNA damage regulated autophagy modulator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018370.2 Intron NP_060840.2
XM_005269004.2 Intron XP_005269061.1
XM_005269005.2 Intron XP_005269062.1
XM_017019578.1 Intron XP_016875067.1

View Full Product Details