Product Details
- SNP ID
-
rs146477429
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:52395164 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGGCGTGCTGACCCCACATGGCTC[A/G]TACAGGAAGGCGCCTTTGGAGCTGC
- Phenotype
-
MIM: 601078
MIM: 602766
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
KRT82
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1732263] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KRT82
- Gene Name
- keratin 82
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_033033.3 |
1468 |
Silent Mutation |
TAC,TAT |
Y451Y |
NP_149022.3 |
- Gene
- KRT84
- Gene Name
- keratin 84
There are no transcripts associated with this gene.
View Full Product Details