Product Details

SNP ID
rs146499431
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:122729558 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGTAGGTGAAGCTGAGGGTTATG[G/T]GCAGGGCCCCATGGACAGAGGGATC
Phenotype
MIM: 606923
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HCAR1 PubMed Links

Gene Details

Gene
HCAR1
Gene Name
hydroxycarboxylic acid receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032554.3 1025 Missense Mutation CAC,CCC H261P NP_115943.1

View Full Product Details