Product Details
- SNP ID
-
rs146528092
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:6541628 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTGCCGGTTCATGTCGTTCTTGGC[C/T]GTGGCCAACTCCAGCTGTGGTGGGG
- Phenotype
-
MIM: 138400
MIM: 610495
MIM: 615638
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GAPDH
PubMed Links
Gene Details
- Gene
- GAPDH
- Gene Name
- glyceraldehyde-3-phosphate dehydrogenase
There are no transcripts associated with this gene.
- Gene
- IFFO1
- Gene Name
- intermediate filament family orphan 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001039670.2 |
1807 |
Silent Mutation |
ACA,ACG |
T490T |
NP_001034759.1 |
NM_001193457.1 |
1807 |
Silent Mutation |
ACA,ACG |
T498T |
NP_001180386.1 |
NM_080730.4 |
1807 |
Silent Mutation |
ACA,ACG |
T489T |
NP_542768.2 |
XM_006718975.3 |
1807 |
Silent Mutation |
ACA,ACG |
T489T |
XP_006719038.1 |
XM_006718976.3 |
1807 |
Silent Mutation |
ACA,ACG |
T487T |
XP_006719039.1 |
XM_006718977.3 |
1807 |
Silent Mutation |
ACA,ACG |
T486T |
XP_006719040.1 |
XM_006718981.3 |
1807 |
Silent Mutation |
ACA,ACG |
T182T |
XP_006719044.1 |
XM_011520947.2 |
1807 |
Silent Mutation |
ACA,ACG |
T578T |
XP_011519249.1 |
XM_011520948.2 |
1807 |
Silent Mutation |
ACA,ACG |
T577T |
XP_011519250.1 |
XM_011520949.2 |
1807 |
Silent Mutation |
ACA,ACG |
T575T |
XP_011519251.1 |
XM_011520952.2 |
1807 |
Silent Mutation |
ACA,ACG |
T270T |
XP_011519254.1 |
XM_017019169.1 |
1807 |
Intron |
|
|
XP_016874658.1 |
XM_017019170.1 |
1807 |
Silent Mutation |
ACA,ACG |
T515T |
XP_016874659.1 |
XM_017019171.1 |
1807 |
Silent Mutation |
ACA,ACG |
T488T |
XP_016874660.1 |
XM_017019172.1 |
1807 |
Silent Mutation |
ACA,ACG |
T271T |
XP_016874661.1 |
XM_017019173.1 |
1807 |
Silent Mutation |
ACA,ACG |
T268T |
XP_016874662.1 |
- Gene
- NCAPD2
- Gene Name
- non-SMC condensin I complex subunit D2
There are no transcripts associated with this gene.
View Full Product Details