Product Details

SNP ID
rs146528669
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:53051898 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATAGCTTCCGGGAGAAGGTTTTCC[A/G]GAAGAAACCTCCAGTCTGTGCAGTA
Phenotype
MIM: 603928 MIM: 607717
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
EIF4B PubMed Links

Gene Details

Gene
EIF4B
Gene Name
eukaryotic translation initiation factor 4B
There are no transcripts associated with this gene.

Gene
LOC283335
Gene Name
uncharacterized LOC283335
There are no transcripts associated with this gene.

Gene
MIR6757
Gene Name
microRNA 6757
There are no transcripts associated with this gene.

Gene
TNS2
Gene Name
tensin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015319.2 145 Missense Mutation CAG,CGG Q50R NP_056134.2
NM_170754.2 145 Missense Mutation CAG,CGG Q40R NP_736610.2
NM_198316.1 145 UTR 5 NP_938072.1
XM_006719302.3 145 Missense Mutation CAG,CGG Q50R XP_006719365.1
XM_006719303.1 145 Missense Mutation CAG,CGG Q40R XP_006719366.1
XM_011538079.1 145 Missense Mutation CAG,CGG Q34R XP_011536381.1
XM_017019088.1 145 Missense Mutation CAG,CGG Q40R XP_016874577.1
XM_017019089.1 145 Missense Mutation CAG,CGG Q34R XP_016874578.1

View Full Product Details