Product Details

SNP ID
rs146536505
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55681877 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGCTACTTCCCCTACCTGATGGC[C/T]GTGCTGACTCCCAAGAGCAACCGCA
Phenotype
MIM: 600536
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ITGA7 PubMed Links

Gene Details

Gene
ITGA7
Gene Name
integrin subunit alpha 7
There are no transcripts associated with this gene.

Gene
METTL7B
Gene Name
methyltransferase like 7B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152637.2 332 Silent Mutation GCC,GCT A41A NP_689850.2

View Full Product Details