Product Details

SNP ID
rs146563866
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:101280491 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACTGGATGTAGTAAACATGGGTT[A/G]GAGGAGGAAAGAAGTGTGGACAGCA
Phenotype
MIM: 612822
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UTP20 PubMed Links

Gene Details

Gene
UTP20
Gene Name
UTP20, small subunit processome component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014503.2 Intron NP_055318.2

View Full Product Details