Product Details

SNP ID
rs146858292
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:48525807 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGATTCTTGTCTCCATAAGCCCA[G/T]GTATTTCTTCCTGAGTCACCTCTCT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR8S1 PubMed Links

Gene Details

Gene
OR8S1
Gene Name
olfactory receptor family 8 subfamily S member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005203.2 176 Missense Mutation AGG,ATG R59M NP_001005203.2

View Full Product Details