Product Details

SNP ID
rs146954242
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:88148325 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGCTTATAGAAAAGATGGCTAAT[A/G]TTAACCTAAAAGAAATAACCTTAAT
Phenotype
MIM: 610142
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CEP290 PubMed Links

Gene Details

Gene
CEP290
Gene Name
centrosomal protein 290
There are no transcripts associated with this gene.

Gene
TMTC3
Gene Name
transmembrane and tetratricopeptide repeat containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181783.3 224 Missense Mutation ATT,GTT I4V NP_861448.2
XM_011537980.2 224 Missense Mutation ATT,GTT I4V XP_011536282.1
XM_011537981.2 224 Intron XP_011536283.1

View Full Product Details