Product Details

SNP ID
rs147143603
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:43797051 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTCTTGCAGCTAGAATACAGCAT[C/T]CGCTCTCTTATACTGCATGTGTATC
Phenotype
MIM: 606883 MIM: 610932
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IRAK4 PubMed Links

Gene Details

Gene
IRAK4
Gene Name
interleukin 1 receptor associated kinase 4
There are no transcripts associated with this gene.

Gene
TMEM117
Gene Name
transmembrane protein 117
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286211.1 Intron NP_001273140.1
NM_001286212.1 Intron NP_001273141.1
NM_001286213.1 Intron NP_001273142.1
NM_032256.2 Intron NP_115632.1
XM_011538831.2 Intron XP_011537133.1
XM_011538832.2 Intron XP_011537134.1
XM_011538833.2 Intron XP_011537135.1
XM_017020022.1 Intron XP_016875511.1
XM_017020023.1 Intron XP_016875512.1
Gene
TWF1
Gene Name
twinfilin actin binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242397.1 Intron NP_001229326.1
NM_002822.4 Intron NP_002813.3

View Full Product Details