Product Details

SNP ID
rs147300461
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55682042 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCACCGGGCTGCAGGGTCACCTG[C/T]CTAGACCCAAATCCCCACTTTGAGA
Phenotype
MIM: 600536
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ITGA7 PubMed Links

Gene Details

Gene
ITGA7
Gene Name
integrin subunit alpha 7
There are no transcripts associated with this gene.

Gene
METTL7B
Gene Name
methyltransferase like 7B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152637.2 497 Silent Mutation TGC,TGT C96C NP_689850.2

View Full Product Details