Product Details
- SNP ID
-
rs147528205
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:65055155 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTCTTCCATGCCAACCTTCTTGACA[C/T]TGGCATTTGTTGGGTTCATGGCAGG
- Phenotype
-
MIM: 605186
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
WIF1
PubMed Links
Gene Details
- Gene
- WIF1
- Gene Name
- WNT inhibitory factor 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_007191.4 |
1356 |
Silent Mutation |
|
|
NP_009122.2 |
View Full Product Details