Product Details

SNP ID
rs147594088
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:112140981 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAGATCTGAAGACTCACCCTGAA[G/T]TTTGTGGGAGAGAGGGGGAGGAAAA
Phenotype
MIM: 613197
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
TRAFD1 PubMed Links

Gene Details

Gene
TRAFD1
Gene Name
TRAF-type zinc finger domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143906.1 486 Missense Mutation GTT,TTT V134F NP_001137378.1
NM_006700.2 486 Missense Mutation GTT,TTT V134F NP_006691.1

View Full Product Details