Product Details

SNP ID
rs147594620
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:110654295 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAGAAAACAGATGGACAGGGAAG[A/C]TTCTCCTCCTATGCTGGCATAGGAG
Phenotype
MIM: 611227 MIM: 609863
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HVCN1 PubMed Links

Gene Details

Gene
HVCN1
Gene Name
hydrogen voltage gated channel 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040107.1 Intron NP_001035196.1
NM_001256413.1 Intron NP_001243342.1
NM_032369.3 Intron NP_115745.2
XM_005253948.2 Intron XP_005254005.1
XM_011538838.1 Intron XP_011537140.1
XM_011538839.2 Intron XP_011537141.1
XM_011538840.2 Intron XP_011537142.1
XM_011538841.2 Intron XP_011537143.1
XM_011538842.2 Intron XP_011537144.1
XM_011538844.2 Intron XP_011537146.1
XM_011538845.1 Intron XP_011537147.1
XM_011538846.2 Intron XP_011537148.1
XM_011538847.1 Intron XP_011537149.1
XM_017020026.1 Intron XP_016875515.1
XM_017020027.1 Intron XP_016875516.1
Gene
TCTN1
Gene Name
tectonic family member 1
There are no transcripts associated with this gene.

View Full Product Details