Product Details

SNP ID
rs147627051
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:15621412 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTCTTCTCATAATTTCTTGTAACT[C/T]GGAGCTGCCACTGCTATCCTGAAAG
Phenotype
MIM: 600206
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EPS8 PubMed Links

Gene Details

Gene
EPS8
Gene Name
epidermal growth factor receptor pathway substrate 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004447.5 2871 Missense Mutation AAG,GAG K792E NP_004438.3
XM_011520605.2 2871 Missense Mutation AAG,GAG K812E XP_011518907.1

View Full Product Details