Product Details

SNP ID
rs147746301
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:121308552 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCTGCCGGAAGAGCATCGCACA[C/T]CGGTTCCTCTCCTGGGTCTTCCCCA
Phenotype
MIM: 606948
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANAPC5 PubMed Links

Gene Details

Gene
ANAPC5
Gene Name
anaphase promoting complex subunit 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001137559.1 2318 Silent Mutation CGA,CGG R620R NP_001131031.1
NM_016237.4 2318 Silent Mutation CGA,CGG R732R NP_057321.2
XM_005253900.3 2318 Silent Mutation CGA,CGG R719R XP_005253957.1
XM_017019423.1 2318 Silent Mutation CGA,CGG R334R XP_016874912.1
XM_017019424.1 2318 Silent Mutation CGA,CGG R334R XP_016874913.1

View Full Product Details