Product Details

SNP ID
rs148139529
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:120438400 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCTCGCATGTGGAAGACTCTCA[C/T]CTTCTTCGTCGCGCTCCCCGGGGTG
Phenotype
MIM: 602072 MIM: 614943
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COX6A1 PubMed Links

Gene Details

Gene
COX6A1
Gene Name
cytochrome c oxidase subunit 6A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004373.3 Intron NP_004364.2
Gene
GATC
Gene Name
glutamyl-tRNA amidotransferase subunit C
There are no transcripts associated with this gene.

Gene
TRIAP1
Gene Name
TP53 regulated inhibitor of apoptosis 1
There are no transcripts associated with this gene.

View Full Product Details