Product Details

SNP ID
rs148151655
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:92424711 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACAAATGCTCCTTTCATTCCTCT[A/G]TTTACAGACCTGCCGCAGACAATTC
Phenotype
MIM: 616988 MIM: 616989
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CLLU1 PubMed Links
Additional Information
For this assay, SNP(s) [rs12580153] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
CLLU1
Gene Name
chronic lymphocytic leukemia up-regulated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025233.2 753 Missense Mutation ATT,GTT I11V NP_001020404.1
Gene
CLLU1OS
Gene Name
chronic lymphocytic leukemia up-regulated 1 opposite strand
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025232.1 753 Intron NP_001020403.1
XM_017019673.1 753 Intron XP_016875162.1

View Full Product Details