Product Details

SNP ID
rs148282251
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:1946279 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCATAGATTATATTTAAGAAGTTG[A/T]CATCATTCTGGAAAACAAATCGAAA
Phenotype
MIM: 609843
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
DCP1B PubMed Links

Gene Details

Gene
DCP1B
Gene Name
decapping mRNA 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319292.1 2006 Intron NP_001306221.1
NM_152640.4 2006 Missense Mutation GAC,GTC D594V NP_689853.3
XM_011520927.2 2006 Missense Mutation GAC,GTC D492V XP_011519229.1
Gene
LINC00940
Gene Name
long intergenic non-protein coding RNA 940
There are no transcripts associated with this gene.

View Full Product Details