Product Details
- SNP ID
-
rs148570766
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:108788025 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATCCACGAAGGGCTCTTTAAGTTT[C/T]CTGGGGCGGGTTTCCCTGATGGTTT
- Phenotype
-
MIM: 606778
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SSH1
PubMed Links
Gene Details
- Gene
- SSH1
- Gene Name
- slingshot protein phosphatase 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001161330.1 |
2997 |
Intron |
|
|
NP_001154802.1 |
NM_001161331.1 |
2997 |
Intron |
|
|
NP_001154803.1 |
NM_018984.3 |
2997 |
Missense Mutation |
GAA,GGA |
E1038G |
NP_061857.3 |
XM_005268984.4 |
2997 |
Missense Mutation |
GAA,GGA |
E1038G |
XP_005269041.1 |
XM_005268985.2 |
2997 |
Missense Mutation |
GAA,GGA |
E958G |
XP_005269042.1 |
XM_011538497.1 |
2997 |
Missense Mutation |
GAA,GGA |
E1049G |
XP_011536799.1 |
XM_011538499.1 |
2997 |
Missense Mutation |
GAA,GGA |
E982G |
XP_011536801.1 |
XM_011538500.2 |
2997 |
Missense Mutation |
GAA,GGA |
E958G |
XP_011536802.1 |
XM_011538501.2 |
2997 |
Missense Mutation |
GAA,GGA |
E958G |
XP_011536803.1 |
XM_017019491.1 |
2997 |
Missense Mutation |
GAA,GGA |
E1022G |
XP_016874980.1 |
XM_017019492.1 |
2997 |
Missense Mutation |
GAA,GGA |
E912G |
XP_016874981.1 |
XM_017019493.1 |
2997 |
Missense Mutation |
GAA,GGA |
E726G |
XP_016874982.1 |
XM_017019494.1 |
2997 |
Missense Mutation |
GAA,GGA |
E726G |
XP_016874983.1 |
View Full Product Details