Product Details
- SNP ID
-
rs148737790
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:68155458 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCATCACTTGGATGAGTTCATGTAT[C/T]GCTTTGCGTTGGACATTCAAGTCAG
- Phenotype
-
MIM: 147570
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
IFNG
PubMed Links
Gene Details
- Gene
- IFNG
- Gene Name
- interferon gamma
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000619.2 |
522 |
Silent Mutation |
GCA,GCG |
A132A |
NP_000610.2 |
View Full Product Details