Product Details

SNP ID
rs148781361
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:53327088 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATAACCATGGCAACAGGGGATTAA[C/T]CTGATGGGGTCATGGTGTCTAAGGG
Phenotype
MIM: 605378 MIM: 606633
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AAAS PubMed Links

Gene Details

Gene
AAAS
Gene Name
aladin WD repeat nucleoporin
There are no transcripts associated with this gene.

Gene
SP7
Gene Name
Sp7 transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001173467.2 2677 UTR 3 NP_001166938.1
NM_001300837.1 2677 UTR 3 NP_001287766.1
NM_152860.1 2677 UTR 3 NP_690599.1
XM_011537900.2 2677 UTR 3 XP_011536202.1

View Full Product Details