Product Details

SNP ID
rs149081968
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:112140995 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCACCCTGAAGTTTGTGGGAGAGA[C/G]GGGGAGGAAAAGAGAAATGAGGTTG
Phenotype
MIM: 613197
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TRAFD1 PubMed Links

Gene Details

Gene
TRAFD1
Gene Name
TRAF-type zinc finger domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143906.1 500 Silent Mutation GAC,GAG D138E NP_001137378.1
NM_006700.2 500 Silent Mutation GAC,GAG D138E NP_006691.1

View Full Product Details